accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1196-38-9

IUPAC Name: 1,2,3,4-tetrahydroisoquinolin-1-one
Synonyms: 3,4-dihydro-2H-isoquinolin-1-one
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: YWPMKTWUFVOFPL-UHFFFAOYSA-N
SMILES: O=C1NCCC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results