accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1197334-04-5

H3CNNOHNNHCl

CAS RN 1197334-04-5

IUPAC Name: 5-methyl-N-(pyridin-3-yl)-1H,2H,3H,5H-pyrrolo[2,3-f]indole-1-carboxamide hydrochloride
Synonyms: 5-methyl-N-(pyridin-3-yl)-2H,3H-pyrrolo[2,3-f]indole-1-carboxamide hydrochloride
Molecular Weight (g/mol): 328.8
Molecular Formula: C17H17ClN4O
InChi Key: VGEMBOFBPSNOIO-UHFFFAOYSA-N
SMILES: Cl.CN1C=CC2=CC3=C(CCN3C(=O)NC3=CN=CC=C3)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results