accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COSiOFFFFFOH3COCH3

CAS RN 1197981-13-7

IUPAC Name: triethoxy[11-(2,3,4,5,6-pentafluorophenoxy)undecyl]silane
Synonyms: triethoxy[11-(2,3,4,5,6-pentafluorophenoxy)undecyl]silane
Molecular Weight (g/mol): 500.62
Molecular Formula: C23H37F5O4Si
InChi Key: YAAQJDPGHMLLJH-UHFFFAOYSA-N
SMILES: CCO[Si](CCCCCCCCCCCOC1=C(F)C(F)=C(F)C(F)=C1F)(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results