accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 119851-23-9

OHOOOOOOHMg

CAS RN 119851-23-9

IUPAC Name: magnesium(2+) 2-(carboxymethyl)-2-hydroxybutanedioate hydrate
Synonyms: magnesium(2+) manganese citrate hydrate
Molecular Weight (g/mol): 232.43
Molecular Formula: C6H8MgO8
InChi Key: HSXLIRULMXTTLM-UHFFFAOYNA-L
SMILES: O.[Mg++].OC(=O)CC(O)(CC([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results