accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFOOO

CAS RN 119895-68-0

IUPAC Name: 2,2-difluoro-2H-1,3-benzodioxole-4-carbaldehyde
Synonyms: 2,2-difluoro-1,3-benzodioxole-4-carbaldehyde
Molecular Weight (g/mol): 186.11
Molecular Formula: C8H4F2O3
InChi Key: NIBFJPXGNVPNHK-UHFFFAOYSA-N
SMILES: FC1(F)OC2=CC=CC(C=O)=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results