accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1199-01-5

IUPAC Name: 2-phenyl-4,5-dihydro-1,3-oxazol-5-one
Synonyms: phenyloxazolone 2-phenyloxazolone
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: QKCKCXFWENOGER-UHFFFAOYSA-N
SMILES: O=C1CN=C(O1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results