accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1199-77-5

IUPAC Name: (2E)-2-methyl-3-phenylprop-2-enoic acid
Synonyms: α-methylcinnamic acid
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: XNCRUNXWPDJHGV-BQYQJAHWSA-N
SMILES: C\C(=C/C1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results