accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 120-15-0

IUPAC Name: 6-methoxy-1,2,3,4-tetrahydroquinoline
Synonyms: 6-methoxy-1,2,3,4-tetrahydroquinoline
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: FRXSZNDVFUDTIR-UHFFFAOYSA-N
SMILES: COC1=CC=C2NCCCC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results