accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 120-23-0

IUPAC Name: 2-(naphthalen-2-yloxy)acetic acid
Synonyms: NOXA betapal 2-noxa gerlach 1396 (2-naphthloxy)acetic acid acide naphthyloxyacetique 2-naphthoxyacetic acid β-naphthoxyacetic acid acide naphtyloxyacetique β-naphthyloxyacetic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C12H10O3
InChi Key: RZCJYMOBWVJQGV-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results