accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 120-43-4

IUPAC Name: ethyl piperazine-1-carboxylate
Synonyms: ethyl piperazine-1-carboxylate
Molecular Weight (g/mol): 158.2
Molecular Formula: C7H14N2O2
InChi Key: LNOQURRKNJKKBU-UHFFFAOYSA-N
SMILES: CCOC(=O)N1CCNCC1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)