accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHClCl

CAS RN 120-83-2

IUPAC Name: 2,4-dichlorophenol
Synonyms: 2,4-dichlorophenol ccris 657 rcra waste number U081
Molecular Weight (g/mol): 163
Molecular Formula: C6H4Cl2O
InChi Key: HFZWRUODUSTPEG-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (6)
Solvent or Matrix 
  • (1)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
Concentration 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)