accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3N

CAS RN 120-94-5

IUPAC Name: 1-methylpyrrolidine
Synonyms: 1-methyl-pyrrolidine
Molecular Weight (g/mol): 85.15
Molecular Formula: C5H11N
InChi Key: AVFZOVWCLRSYKC-UHFFFAOYSA-N
SMILES: CN1CCCC1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)