accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrO

CAS RN 1200-03-9

IUPAC Name: (4-bromobutoxy)benzene
Synonyms: (4-bromobutoxy)benzene
Molecular Weight (g/mol): 229.12
Molecular Formula: C10H13BrO
InChi Key: QBLISOIWPZSVIK-UHFFFAOYSA-N
SMILES: BrCCCCOC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results