accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1200-07-3

IUPAC Name: (2E)-3-(4-bromophenyl)prop-2-enoic acid
Synonyms: (2E)-3-(4-bromophenyl)prop-2-enoic acid
Molecular Weight (g/mol): 227.06
Molecular Formula: C9H7BrO2
InChi Key: CPDDDTNAMBSPRN-ZZXKWVIFSA-N
SMILES: OC(=O)\C=C\C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results