accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12003-65-5

OOOAlLa

CAS RN 12003-65-5

IUPAC Name: aluminium(3+) lanthanum(3+) trioxidandiide
Synonyms: aluminium(3+) lanthanum(3+) trioxidandiide
Molecular Weight (g/mol): 213.88
Molecular Formula: AlLaO3
InChi Key: BOIGHUSRADNYQR-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Al+3].[La+3]
Physical Form 
  • (6)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (6)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)