accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12004-83-0

AlAlAlZr

CAS RN 12004-83-0

IUPAC Name: trialuminium zirconium
Synonyms: trialuminium zirconium
Molecular Weight (g/mol): 172.17
Molecular Formula: Al3Zr
InChi Key: PGLXYLYDLCVKEE-UHFFFAOYSA-N
SMILES: [Al].[Al].[Al].[Zr]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results