accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12005-82-2

FAsFFFFFAg

CAS RN 12005-82-2

IUPAC Name: silver(1+) hexafluoro-λ⁵-arsanuide
Synonyms: silver(1+) hexafluoro-λ⁵-arsanuide
Molecular Weight (g/mol): 296.78
Molecular Formula: AgAsF6
InChi Key: UNSKNPXMHZPWPB-UHFFFAOYSA-N
SMILES: [Ag+].F[As-](F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results