accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

AsCdAsCdCd

CAS RN 12006-15-4

IUPAC Name: bis(cadmio-λ²-arsanyl) cadmium
Synonyms: cadmium arsenide
Molecular Weight (g/mol): 487.09
Molecular Formula: As2Cd3
InChi Key: FSIONULHYUVFFA-UHFFFAOYSA-N
SMILES: [Cd].[As]=[Cd].[As]=[Cd]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results