accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BMo

CAS RN 12006-98-3

IUPAC Name: molybdenioborane
Synonyms: molybdenioborane
Molecular Weight (g/mol): 106.76
Molecular Formula: BMo
InChi Key: LIGIGBHZRAABEF-UHFFFAOYSA-N
SMILES: [B+]=[Mo-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results