accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12007-29-3

BNb

CAS RN 12007-29-3

IUPAC Name: niobioborane
Synonyms: niobioborane
Molecular Weight (g/mol): 103.72
Molecular Formula: BNb
InChi Key: VDZMENNHPJNJPP-UHFFFAOYSA-N
SMILES: B#[Nb]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results