accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BBBBBBGd

CAS RN 12008-06-9

IUPAC Name: gadolinium(2+) pentacyclo[3.1.0.0¹,³.0²,⁴.0⁴,⁶]hexaborane-1,4-diuide
Synonyms: gadolinium(2+) pentacyclo[3.1.0.0¹,³.0²,⁴.0⁴,⁶]hexaborane-1,4-diuide
Molecular Weight (g/mol): 222.11
Molecular Formula: B6Gd
InChi Key: FYKPSYVUFHLGOY-UHFFFAOYSA-N
SMILES: [Gd++].B12B3[B-]11B4B1[B-]234

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results