accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 120107-61-1

IUPAC Name: imidazo[2,1-b][1,3]thiazole-6-carbaldehyde
Synonyms: imidazo[2,1-b][1,3]thiazole-6-carbaldehyde
Molecular Weight (g/mol): 152.17
Molecular Formula: C6H4N2OS
InChi Key: VYBDPAAWKNKEKY-UHFFFAOYSA-N
SMILES: O=CC1=CN2C=CSC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results