accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12012-35-0

CrCrCr

CAS RN 12012-35-0

IUPAC Name: λ²-chromium(2+) bis(chromiomethanide)
Synonyms: chromium carbide
Molecular Weight (g/mol): 180.01
Molecular Formula: C2Cr3
InChi Key: GVEHJMMRQRRJPM-UHFFFAOYSA-N
SMILES: [Cr++].[C-]#[Cr].[C-]#[Cr]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results