accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12012-95-2

IUPAC Name: bis(chloropalladiumylium); bis(prop-2-en-1-ide)
Synonyms: allylpalladium chloride dimer
Molecular Weight (g/mol): 365.89
Molecular Formula: C6H10Cl2Pd2
InChi Key: PENAXHPKEVTBLF-UHFFFAOYSA-L
SMILES: Cl[Pd+].Cl[Pd+].[CH2-]C=C.[CH2-]C=C
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)