accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CH3CCH3H3CNOCH3

CAS RN 1201644-36-1

IUPAC Name: 2-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Synonyms: 2-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Molecular Weight (g/mol): 285.15
Molecular Formula: C16H20BNO3
InChi Key: DVGWZOHNHYZTKK-UHFFFAOYSA-N
SMILES: COC1=NC2=CC=C(C=C2C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results