accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12017-12-8

SiCoSi

CAS RN 12017-12-8

IUPAC Name: (silanylidenecobaltio)silane
Synonyms: (silanylidenecobaltio)silane
Molecular Weight (g/mol): 115.1
Molecular Formula: CoSi2
InChi Key: QKWVWIGBHQUEHX-UHFFFAOYSA-N
SMILES: [Si]=[Co]=[Si]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results