accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOOFeFeSrSr

CAS RN 12023-91-5

IUPAC Name: diiron(3+) distrontium(2+) pentaoxidandiide
Synonyms: diiron(3+) distrontium(2+) pentaoxidandiide
Molecular Weight (g/mol): 366.93
Molecular Formula: Fe2O5Sr2
InChi Key: GKHGYOYJUWWDJG-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[Fe+3].[Fe+3].[Sr++].[Sr++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results