CAS RN 12025-39-7
IUPAC Name:
germanium tellurium
Synonyms:
germanium telluride
Molecular Weight (g/mol):
200.23
Molecular Formula:
GeTe
InChi Key:
VDDXNVZUVZULMR-UHFFFAOYSA-N
SMILES:
[Ge].[Te]
1
–
1
of
1
results
1
Germanium(II) telluride, 99.999%
CAS: 12025-39-7 Molecular Formula: GeH2Te Molecular Weight (g/mol): 202.25 MDL Number: MFCD00135541 InChI Key: SBMPISTWTKUJKL-UHFFFAOYSA-N Synonym: germanium telluride,germanium tel-luride,tellanylidenegermanylidene,germanium telluride trace metals basis PubChem CID: 16213264 IUPAC Name: germanetellone SMILES: [GeH2]=[Te]