CAS RN 12031-80-0
IUPAC Name:
dilithium(1+) dioxidanediide
Synonyms:
lithium peroxide
Molecular Weight (g/mol):
45.88
Molecular Formula:
Li2O2
InChi Key:
HPGPEWYJWRWDTP-UHFFFAOYSA-N
SMILES:
[Li+].[Li+].[O-][O-]
1
–
2
of
2
results
1
Lithium peroxide, 95%, Thermo Scientific™
CAS: 12031-80-0 Molecular Formula: Li2O2 Molecular Weight (g/mol): 45.88 MDL Number: MFCD00016184 InChI Key: HPGPEWYJWRWDTP-UHFFFAOYSA-N Synonym: lithium peroxide li2 o2,lithium peroxide un1472 oxidizer,lithium oxide,dilithium peroxide,unii-9anx556r5f PubChem CID: 25489 IUPAC Name: dilithium;peroxide SMILES: [Li+].[Li+].[O-][O-]
2
Lithium peroxide, 95%
CAS: 12031-80-0 Molecular Formula: Li2O2 MDL Number: MFCD00016184 InChI Key: HPGPEWYJWRWDTP-UHFFFAOYSA-N PubChem CID: 25489 IUPAC Name: dilithium;peroxide SMILES: [Li+].[Li+].[O-][O-]