accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12032-20-1

IUPAC Name: dilutetium(3+) trioxidandiide
Synonyms: lutetium(III) oxide
Molecular Weight (g/mol): 397.93
Molecular Formula: Lu2O3
InChi Key: UGBIHFMRUDAMBY-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Lu+3].[Lu+3]
Physical Form 
  • (9)
  • (3)
Quantity 
  • (5)
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (4)
  • (1)
  • (2)
Grade 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results