accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12033-89-5

NNSiNNSiSi

CAS RN 12033-89-5

IUPAC Name: 1Si-hexacyclo[3.1.1.0¹,⁴.0²,⁵.0³,⁶.0³,⁷]trisilazane
Synonyms: silicon nitride
Molecular Weight (g/mol): 140.28
Molecular Formula: N4Si3
InChi Key: HQVNEWCFYHHQES-UHFFFAOYSA-N
SMILES: N12[Si]34N5[Si]11N3[Si]25N41
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
Physical Form 
  • (14)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (14)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (14)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)