accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12034-59-2

ONbO

CAS RN 12034-59-2

IUPAC Name: dioxoniobium
Synonyms: niobium dioxide
Molecular Weight (g/mol): 124.9
Molecular Formula: NbO2
InChi Key: HFLAMWCKUFHSAZ-UHFFFAOYSA-N
SMILES: O=[Nb]=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results