accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12035-72-2

SNiNiNiS

CAS RN 12035-72-2

IUPAC Name: (trinickel-1-ylidene)-1λ⁴-disulfene
Synonyms: trinickel-1-ylidene-1λ⁴-disulfene
Molecular Weight (g/mol): 240.2
Molecular Formula: Ni3S2
InChi Key: UPSUSTJZZWSTPN-UHFFFAOYSA-N
SMILES: [Ni][Ni][Ni]=S=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results