accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12036-32-7

IUPAC Name: dipraseodymium(3+) trioxidandiide
Synonyms: praseodymium(III) oxide
Molecular Weight (g/mol): 329.81
Molecular Formula: O3Pr2
InChi Key: MMKQUGHLEMYQSG-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Pr+3].[Pr+3]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results