accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12036-35-0

OOORhRh

CAS RN 12036-35-0

IUPAC Name: dirhodium(3+) trioxidandiide
Synonyms: rhodium(III) oxide
Molecular Weight (g/mol): 253.81
Molecular Formula: O3Rh2
InChi Key: XBBXDTCPEWHXKL-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Rh+3].[Rh+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results