accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12042-68-1

CaOAlOOAlO

CAS RN 12042-68-1

IUPAC Name: calcium dialumanoylolate
Synonyms: calcium dialumanoylolate
Molecular Weight (g/mol): 158.04
Molecular Formula: Al2CaO4
InChi Key: XFWJKVMFIVXPKK-UHFFFAOYSA-N
SMILES: [Ca++].[O-][Al]=O.[O-][Al]=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results