accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12045-63-5

BBTi

CAS RN 12045-63-5

IUPAC Name: diboron titanium
Synonyms: titanium diboride
Molecular Weight (g/mol): 69.49
Molecular Formula: B2Ti
InChi Key: QYEXBYZXHDUPRC-UHFFFAOYSA-N
SMILES: [B].[B].[Ti]
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results