accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12045-78-2

H2OH2OH2OH2OKOBOO

CAS RN 12045-78-2

IUPAC Name: potassium tetrahydrate borate
Synonyms: potassium tetrahydrate borate
Molecular Weight (g/mol): 169.97
Molecular Formula: BH8KO7
InChi Key: SAAWPUYPXWYTFF-UHFFFAOYSA-N
SMILES: O.O.O.O.[K+].[O-]B([O-])[O-]
Physical Form 
  • (1)
  • (2)
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)