accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SOOClOO

CAS RN 1205-30-7

IUPAC Name: 4-chloro-3-sulfamoylbenzoate
Synonyms: 4-chloro-3-sulfamoylbenzoate
Molecular Weight (g/mol): 234.63
Molecular Formula: C7H5ClNO4S
InChi Key: FHQAWINGVCDTTG-UHFFFAOYSA-M
SMILES: NS(=O)(=O)C1=CC(=CC=C1Cl)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results