accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH

CAS RN 1205-64-7

IUPAC Name: 3-methyl-N-phenylaniline
Synonyms: 3-methyl diphenyl amine
Molecular Weight (g/mol): 183.25
Molecular Formula: C13H13N
InChi Key: TWPMMLHBHPYSMT-UHFFFAOYSA-N
SMILES: CC1=CC=CC(NC2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results