CAS RN 12054-48-7
IUPAC Name:
nickel(2+) bis(λ³-oxidanide)
Synonyms:
nickel(II) hydroxide
Molecular Weight (g/mol):
94.72
Molecular Formula:
H4NiO2
InChi Key:
RDZMIMIDOVRADK-UHFFFAOYSA-N
SMILES:
[OH2-].[OH2-].[Ni++]
1
–
2
of
2
results
1
Nickel(II) hydroxide, for analysis
CAS: 12054-48-7 Molecular Formula: H2NiO2 Molecular Weight (g/mol): 92.71 MDL Number: MFCD00011140 InChI Key: BFDHFSHZJLFAMC-UHFFFAOYSA-L IUPAC Name: nickel(2+) dihydroxide SMILES: [OH-].[OH-].[Ni++]
2
Nickel(II) hydroxide, Ni 61%
CAS: 12054-48-7 Molecular Formula: H2NiO2 Molecular Weight (g/mol): 92.71 MDL Number: MFCD00011140 InChI Key: BFDHFSHZJLFAMC-UHFFFAOYSA-L IUPAC Name: nickel(2+) dihydroxide SMILES: [OH-].[OH-].[Ni++]