CAS RN 12054-85-2
Ammonium molybdate(VI) tetrahydrate, 99+%, for analysis
CAS: 12054-85-2 Molecular Formula: H24Mo7N6O24·4H2O Molecular Weight (g/mol): 1235.86
Ammonium molybdate(VI) tetrahydrate, ACS reagent
CAS: 12054-85-2 Molecular Formula: H24Mo7N6O24·4H2O Molecular Weight (g/mol): 1235.86 MDL Number: MFCD00167059
Ammonium Molybdate Tetrahydrate, Certified AR for Analysis, Fisher Chemical™
CAS: 12054-85-2 Molecular Formula: H6MoNO2 Molecular Weight (g/mol): 148.00 MDL Number: 167059 InChI Key: XCOMAVCHADVANQ-UHFFFAOYSA-O IUPAC Name: ammonium hydrate molybdenum oxidandiide SMILES: [NH4+].O.[O--].[Mo]
Ammonium molybdate (para) tetrahydrate, 99%
CAS: 12054-85-2 Molecular Formula: (NH4)6Mo7O24·4H2O MDL Number: MFCD00167059 Synonym: Ammonium heptamolybdate tetrahydrate; Ammonium paramolybdate tetrahydrate
Ammonium molybdate (para) tetrahydrate, ACS, 81-83% as MoO{3}
CAS: 12054-85-2 Molecular Formula: H24Mo7N6O24·4H2O MDL Number: MFCD00167059 Synonym: Ammonium paramolybdate
Ammonium molybdate tetrahydrate, For ACS analysis, 81 to 83% MoO3, MP Biomedicals™
CAS: 12054-85-2 Molecular Formula: (NH4)6 Mo7O24·4H2O Synonym: Molybdic acid ammonium salt tetrahydrate
Ammonium molybdate tetrahydrate, ACS Reagent, Solstice
CAS: 12054-85-2 MDL Number: MFCD00167059
Ammonium Molybdate Tetrahydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Ammonium Molybdate Tetrahydrate, puriss. p.a. ACS Reagent, Solstice
CAS: 12054-85-2 MDL Number: MFCD00167059
Ammonium Molybdate Tetrahydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Ammonium Molybdate Tetrahydrate, Analytical Grade, Penta Chemicals
CAS: 12054-85-2 Molecular Weight (g/mol): 1235.86
- 1
- 2