CAS RN 1206593-32-9
IUPAC Name:
4-(3-chloro-2-fluorophenoxy)-3-fluoroaniline
Synonyms:
4-(3-chloro-2-fluorophenoxy)-3-fluoroaniline
Molecular Weight (g/mol):
255.65
Molecular Formula:
C12H8ClF2NO
InChi Key:
VLAMCDVNMXJZMN-UHFFFAOYSA-N
SMILES:
NC1=CC(F)=C(OC2=C(F)C(Cl)=CC=C2)C=C1
1
–
1
of
1
results
1
4-(3-Chloro-2-fluorophenoxy)-3-fluoroaniline, 97%, Thermo Scientific™
CAS: 1206593-32-9 Molecular Formula: C12H8ClF2NO Molecular Weight (g/mol): 255.649 MDL Number: MFCD13196577 InChI Key: VLAMCDVNMXJZMN-UHFFFAOYSA-N Synonym: 4-3-chloro-2-fluorophenoxy-3-fluoroaniline,3-fluoro-4-3-chloro-2-fluorophenoxy aniline,4-3-chloro-2-fluorophenoxy-3-fluorophenylamine PubChem CID: 57365060 IUPAC Name: 4-(3-chloro-2-fluorophenoxy)-3-fluoroaniline SMILES: C1=CC(=C(C(=C1)Cl)F)OC2=C(C=C(C=C2)N)F