CAS RN 1206593-35-2
IUPAC Name:
2-[4-ethoxy-3-(trifluoromethyl)phenyl]acetonitrile
Synonyms:
2-[4-ethoxy-3-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol):
229.2
Molecular Formula:
C11H10F3NO
InChi Key:
DMJJFPFYFKGPBW-UHFFFAOYSA-N
SMILES:
CCOC1=C(C=C(CC#N)C=C1)C(F)(F)F
1
–
1
of
1
results
1
4-Ethoxy-3-(trifluoromethyl)phenylacetonitrile, 97%, Thermo Scientific™
CAS: 1206593-35-2 Molecular Formula: C11H10F3NO Molecular Weight (g/mol): 229.202 MDL Number: MFCD13196585 InChI Key: DMJJFPFYFKGPBW-UHFFFAOYSA-N Synonym: 4-ethoxy-3-trifluoromethyl phenylacetonitrile,2-4-ethoxy-3-trifluoromethyl phenyl acetonitrile,2-4-ethoxy-3-trifluoromethyl phenyl ethanenitrile PubChem CID: 57361408 IUPAC Name: 2-[4-ethoxy-3-(trifluoromethyl)phenyl]acetonitrile SMILES: CCOC1=C(C=C(C=C1)CC#N)C(F)(F)F