accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12067-57-1

SiSiSiTiTiTiTiTi

CAS RN 12067-57-1

IUPAC Name: trisilicon pentatitanium
Synonyms: trisilicon pentatitanium
Molecular Weight (g/mol): 323.59
Molecular Formula: Si3Ti5
InChi Key: GNKTZDSRQHMHLZ-UHFFFAOYSA-N
SMILES: [Si].[Si].[Si].[Ti].[Ti].[Ti].[Ti].[Ti]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results