accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12069-69-1

OHOHCuCuOOO

CAS RN 12069-69-1

IUPAC Name: dicopper(2+) dihydroxide carbonate
Synonyms: basic copper carbonate
Molecular Weight (g/mol): 221.11
Molecular Formula: CH2Cu2O5
InChi Key: ZMMDPCMYTCRWFF-UHFFFAOYSA-J
SMILES: [OH-].[OH-].[Cu++].[Cu++].[O-]C([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (6)
Physical Form 
  • (3)
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Grade 
  • (1)
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (6)
  • (3)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)