accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1207-15-4

IUPAC Name: 4,12-dimethyl-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms: 4,12-dimethyl-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol): 212.31
Molecular Formula: C14H12S
InChi Key: RRYWCJRYULRSJM-UHFFFAOYSA-N
SMILES: CC1=CC2=C(SC3=C2C=C(C)C=C3)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results