accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12070-14-3

ZrC

CAS RN 12070-14-3

IUPAC Name: methane zirconium
Synonyms: zirconium carbide
Molecular Weight (g/mol): 107.27
Molecular Formula: CH4Zr
InChi Key: UGHSGZIDZZRZKT-UHFFFAOYSA-N
SMILES: C.[Zr]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results