accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 120711-81-1

OHNNOO

CAS RN 120711-81-1

IUPAC Name: 7-nitro-3,4-dihydro-2H-1,4-benzoxazine
Synonyms: 7-nitro-3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol): 180.16
Molecular Formula: C8H8N2O3
InChi Key: YKCFDUNYLMTXFC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=C(NCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results