CAS RN 1207456-01-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(8S,9R)-BMN 673, TRC
CAS: 1207456-01-6 Molecular Formula: C19H14F2N6O Molecular Weight (g/mol): 380.35 Synonym: (8S,9R)-BMN 673,(8S,9R)-5-Fluoro-8-(4-fluorophenyl)-2,7,8,9-tetrahydro-9-(1-methyl-1H-1,2,4-triazol-5-yl)-3H-pyrido[4,3,2-de]phthalazin-3-one SMILES: Cn1ncnc1[C@@H]2[C@H](Nc3cc(F)cc4C(=O)NN=C2c34)c5ccc(F)cc5